Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dehydroisoandrosterone, 99%, Thermo Scientific Chemicals

Catalog No. AC154980100
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154980100 10 g Glass bottle
AC154980025 2.5 g Glass bottle
AC154980500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC154980100 Supplier Thermo Scientific Chemicals Supplier No. 154980100
Only null left
Add to Cart

(+)-Dehydroisoandrosterone, CAS # 53-43-0, also known as androstenolone, is an endogenous steroid hormone precursor. It is an intermediate in the conversion of cholesterol to androgens and estrogens.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • (+)-Dehydroisoandrosterone is an endogenous steroid hormone precursor that acts as an intermediate in the conversion of cholesterol to androgens and estrogens

Applications

  • It is an endogenous and naturally occurring steroid hormone produced largely by the adrenal glands, but also by the testes and ovaries
  • It has been described that (+)-Dehydroisoandrosterone could have a role in the immune and stress response
  • (+)-Dehydroisoandrosterone can be converted to testosterone, androstenedione, estradiol, and estrone
  • (+)-Dehydroisoandrosterone is derived from cholesterol through the action of two cytochrome P450 enzymes
  • It can induce both direct and indirect in vivo biological effects, by modulating GABA- α receptor and N-Methyl-D-aspartic acid (NMDA) receptor complexes.
  • It can also increase pancreatic β cell insulin secretion and anti-glucocorticoid activity
  • It is useful to detect excess adrenal activity since it is produced nearly exclusively by the adrenal glands
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-43-0
Molecular Formula C19H28O2
Molecular Weight (g/mol) 288.42
MDL Number MFCD00003613
InChI Key FMGSKLZLMKYGDP-USOAJAOKSA-N
Synonym dehydroepiandrosterone, dhea, prasterone, dehydroisoandrosterone, androstenolone, trans-dehydroandrosterone, 3beta-hydroxyandrost-5-en-17-one, diandron, diandrone, psicosterone
PubChem CID 5881
ChEBI CHEBI:28689
IUPAC Name (3S,8R,9S,10R,13S,14S)-3-hydroxy-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-one
SMILES CC12CCC3C(C1CCC2=O)CC=C4C3(CCC(C4)O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 146.0°C to 151.0°C
Color White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 08, IV, 994
Merck Index 15, 2875
Specific Rotation + 12.00
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in benzene,alcohol and diethyl ether,sparingly soluble in chloroform and petrol. ether
Formula Weight 288.42
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material (+)-Dehydroisoandrosterone
Show More Show Less
EINECSNumber 200-175-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.