Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Delta Np73 Antibody (38C674.2), Biotin, Novus Biologicals™
SDP

Catalog No. NBP224873B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224873B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224873B Supplier Novus Biologicals Supplier No. NBP224873B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Delta Np73 Monoclonal specifically detects Delta Np73 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Delta Np73
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 38C674.2
Conjugate Biotin
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. O15350
Gene Alias p53-related protein, p73, tumor protein p73
Gene Symbols p73
Host Species Mouse
Immunogen This antibody was developed against a peptide corresponding to a portion of amino acid residues 1-50 of human dNp73. Mouse and human sequences are more than 95 % identical at these amino acid residues. In mouse, alanine at position 8 is replaced by methionine.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7161
Test Specificity This antibody is reactive to Human, Mouse and Rat.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.