Learn More
Description
Specifications
Specifications
| Antigen | Delta Np73 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay |
| Classification | Monoclonal |
| Clone | DNp73-1.1 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1 ug/ml, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional |
| Formulation | PBS |
| Gene Alias | p53-related protein, p73, tumor protein p73 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
