Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Delta Np73 Antibody (DNp73-1.1), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB388926 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388926 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388926 Supplier Novus Biologicals Supplier No. NBP311702G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Delta Np73 Monoclonal antibody specifically detects Delta Np73 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen Delta Np73
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone DNp73-1.1
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias p53-related protein, p73, tumor protein p73
Host Species Mouse
Immunogen KLH-coupled synthetic peptide MLYVGDPARHLATC
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7161
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.