Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
delta Opioid R/OPRD1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11079877
Description
delta Opioid R/OPRD1 Polyclonal specifically detects delta Opioid R/OPRD1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
delta Opioid R/OPRD1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
delta opioid receptor 1, delta-type opioid receptor, D-OR-1, DOR-1, opioid receptor, delta 1, OPRD | |
Rabbit | |
40 kDa | |
0.1 mg | |
Cancer, Membrane Trafficking and Chaperones | |
4985 | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
1 mg/mL | |
Western Blot 1:100-1:2000 | |
P41143 | |
OPRD1 | |
Synthetic peptide comprising residues 361-372 of the human DOR-1 protein. Reacts with mouse and rat DOR-1. | |
Affinity Purified | |
RUO | |
Primary | |
Delta Opiod Receptor | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction