Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
delta Opioid R/OPRD1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | delta Opioid R/OPRD1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018032
![]() |
Novus Biologicals
NB11079877SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB11079877
![]() |
Novus Biologicals
NB11079877 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
delta Opioid R/OPRD1 Polyclonal specifically detects delta Opioid R/OPRD1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
delta Opioid R/OPRD1 | |
Western Blot 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
Cancer, Membrane Trafficking and Chaperones | |
PBS with 0.02% Sodium Azide | |
delta opioid receptor 1, delta-type opioid receptor, D-OR-1, DOR-1, opioid receptor, delta 1, OPRD | |
OPRD1 | |
IgG | |
Affinity Purified | |
Delta Opiod Receptor |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse, Rat, Human | |
P41143 | |
4985 | |
Synthetic peptide comprising residues 361-372 of the human DOR-1 protein. Reacts with mouse and rat DOR-1. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
40 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title