Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta Opioid R/OPRD1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018033 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018033 0.025 mg
NB11079879 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018033 Supplier Novus Biologicals Supplier No. NB11079879SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

delta Opioid R/OPRD1 Polyclonal specifically detects delta Opioid R/OPRD1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen delta Opioid R/OPRD1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias delta opioid receptor 1, delta-type opioid receptor, D-OR-1, DOR-1, opioid receptor, delta 1, OPRD
Gene Symbols OPRD1
Host Species Rabbit
Immunogen Synthetic peptide comprising residues 3-17 of the mouse and rat DOR-1 protein.
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 4985
Test Specificity Delta Opiod Receptor
Target Species Mouse, Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.