Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta Opioid R/OPRD1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB344776 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344776 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344776 Supplier Novus Biologicals Supplier No. NB11079877PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

delta Opioid R/OPRD1 Polyclonal antibody specifically detects delta Opioid R/OPRD1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen delta Opioid R/OPRD1
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias delta opioid receptor 1, delta-type opioid receptor, D-OR-1, DOR-1, opioid receptor, delta 1, OPRD
Host Species Rabbit
Immunogen Synthetic peptide comprising residues 361-372 of the human DOR-1 protein. Reacts with mouse and rat DOR-1.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 4985
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.