Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Sarcoglycan Antibody (19G8), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP242662G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242662G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242662G Supplier Novus Biologicals Supplier No. NBP242662G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

delta-Sarcoglycan Monoclonal specifically detects delta-Sarcoglycan in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen delta-Sarcoglycan
Applications Western Blot, ELISA
Classification Monoclonal
Clone 19G8
Conjugate DyLight 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD
Gene Symbols SGCD
Host Species Mouse
Immunogen Recombinant human SGCD (57-289aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6444
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.