Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Sarcoglycan Rabbit anti-Human, Mouse, Clone: 3P3R4, Novus Biologicals™
SDP

Catalog No. NB168643 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168643 100 μg
NB168644 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168643 Supplier Novus Biologicals Supplier No. NBP316776100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

delta-Sarcoglycan Monoclonal antibody specifically detects delta-Sarcoglycan in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen delta-Sarcoglycan
Applications Western Blot
Classification Monoclonal
Clone 3P3R4
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 190-289 of human delta-Sarcoglycan (Q92629).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6444
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.