Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
delta-Sarcoglycan Rabbit anti-Human, Mouse, Clone: 3P3R4, Novus Biologicals™

Rabbit Monoclonal Antibody
$206.50 - $492.50
Specifications
Antigen | delta-Sarcoglycan |
---|---|
Clone | 3P3R4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
delta-Sarcoglycan Monoclonal antibody specifically detects delta-Sarcoglycan in Human, Mouse samples. It is validated for Western BlotSpecifications
delta-Sarcoglycan | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6444 | |
IgG | |
Affinity purified |
3P3R4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD | |
A synthetic peptide corresponding to a sequence within amino acids 190-289 of human delta-Sarcoglycan (Q92629). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title