Learn More
Description
Specifications
Specifications
| Antigen | Dendritic Cell Marker |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 33D1 |
| Conjugate | Biotin |
| Dilution | Immunohistochemistry, Immunohistochemistry-Frozen |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | DCs, Dendritic cell |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody was Dendritic Cell Marker. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
