Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dendritic Cells Antibody (RFD1), Novus Biologicals™
SDP

Catalog No. NB10064607 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064607 0.1 mg
NB017618 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064607 Supplier Novus Biologicals Supplier No. NB10064607
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dendritic Cells Monoclonal specifically detects Dendritic Cells in Human, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Dendritic Cells
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RFD1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Alias DCs, Dendritic cell, FDC, FDCs, Follicular dendritic cell
Host Species Mouse
Immunogen Made to Human DENDRITIC CELLS,B CELLS
Purification Method IgM purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64607 recognizes an epitope within the class II MHC complex. In normal tissue NB100-64607 identifies dendritic cells and a proportion of B lymphocytes. Within the macrophage/dendritic cell populations the epitope seen by RFD1 is co-expressed with RFD7 by a subset of cells that exhibit suppressive activity on T cell stimulation. It will not bind to monocytes or any cells within the granulocyte series.
Target Species Human, Primate, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.