Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus 2 Antibody (3H5.1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064446 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064446 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064446 Supplier Novus Biologicals Supplier No. NB10064446
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dengue Virus 2 Monoclonal antibody specifically detects Dengue Virus 2 in Virus samples. It is validated for Immunocytochemistry, Immunofluorescence.

Specifications

Antigen Dengue Virus 2
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3H5.1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:100-1:400
Host Species Mouse
Immunogen Dengue 2 antigen (New Guinea C).
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Reacts with Dengue virus 2.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.