Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus 2 Envelope Antibody (144), PE, Novus Biologicals™
SDP

Catalog No. NB328488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328488 Supplier Novus Biologicals Supplier No. NBP305828PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dengue Virus 2 Envelope Monoclonal antibody specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Dengue Virus 2 Envelope
Applications Western Blot, ELISA
Classification Monoclonal
Clone 144
Conjugate PE
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias Dengue 2 envelope;Dengue envelope;envelope protein, Envelope protein E
Host Species Mouse
Immunogen Produced in mouse immunized with recombinant Dengue Virus 2 Envelope (Accession#: AAC59274.1; Met578-Gln680).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.