Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus 2 Envelope Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB327493 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327493 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327493 Supplier Novus Biologicals Supplier No. NBP306004MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dengue Virus 2 Envelope Polyclonal antibody specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Dengue Virus 2 Envelope
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Dengue 2 envelope;Dengue envelope;envelope protein, Envelope protein E
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Dengue Virus 2 Envelope (Accession#: AAC59274.1; Phe247-Gly675).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.