Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus Antibody (Dengue 1-11(3)), Biotin, Novus Biologicals™
SDP

Catalog No. NB10065685B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065685B 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065685B Supplier Novus Biologicals Supplier No. NB10065685B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dengue Virus Monoclonal specifically detects Dengue Virus in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Dengue Virus
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone Dengue 1-11(3)
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias DEN 1, DEN1, Dengue hemorrhagic fever, Dengue type 1 virus, DENV 1, DENV1
Host Species Mouse
Immunogen Made to Viral DENGUE VIRUS
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity NB100-65685 recognizes all four dengue virus serotypes (DEN-1, DEN-2, DEN-3 and DEN-4 least strongly) of the genus Flavivirus. NB100-65685 detects an anti-E/envelope specific band of 61kDa under reducing conditions in Western blotting. A weak secondary band of 80kDa may also be apparent.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.