Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus NS5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP242901 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242901 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242901 Supplier Novus Biologicals Supplier No. NBP242901
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects NS5 in Virus samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting

Dengue Virus NS5 Polyclonal specifically detects Dengue Virus NS5 in Viral samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dengue Virus NS5
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:100-1:2000
Formulation PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Alias Dengue NS5, Dengue NS5 protein
Gene Symbols POLY
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of Dengue virus NS5 protein (Dengue virus 2 (strain 16681 PDK 53)). The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1489713
Target Species Virus
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.