Learn More
Description
Specifications
Specifications
| Antigen | DENND1A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | Connecdenn, Connecdenn 1, DENN/MADD domain containing 1A, FAM31ADENN domain-containing protein 1A, FLJ21129, FLJ38464, KIAA1608connecdenn, Protein FAM31A, RP11-230L22.3 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 270-527 of human DENND1A (NP_079096.2). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
