Learn More
United Scientific Supplies Density Cube Sets
Shop All United Scientific Supplies, Inc Products
Click to view available options
Quantity:
10 Cubes
12 Cubes
4 Cubes
Description
- For use in science experiments involving density
- Cube measurements are equal
Specifications
Specifications
Product Type | Density Cubes |
Description | Plastic storage box |
Includes | One each of acrylic, oak, pine, poplar, steel, aluminum, copper, brass, nylon, PVC, lignum vitae, and polypropylene cubes; Plastic storage box |
Quantity | 12 Cubes |
Height (English) | 1 in. |
Width (English) | 1 in. |