Learn More
DENTT Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304013AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This antibody recognizes human DENTT (Differentially Expressed Nucleolar TGF-Beta1 Target) protein, a gene product expressed in several normal tissues with the highest level of expression in brain. DENTT is involved in responses mediated by TGF-Beta1 and has distinct domains for direction to different subnuclear locations.Specifications
DENTT | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9H2G4 | |
TSPYL2 | |
Between 643 and 693 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TSPYL2 | |
RP1-290F12.2, CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64061 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.