Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DENTT Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | DENTT |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-6680
![]() |
Bethyl Laboratories, Inc
A304013AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
This antibody recognizes human DENTT (Differentially Expressed Nucleolar TGF-Beta1 Target) protein, a gene product expressed in several normal tissues with the highest level of expression in brain. DENTT is involved in responses mediated by TGF-Beta1 and has distinct domains for direction to different subnuclear locations.Specifications
DENTT | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
RP1-290F12.2, CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX | |
TSPYL2 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9H2G4 | |
64061 | |
Between 643 and 693 | |
Primary | |
4° C | |
TSPYL2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title