Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Deoxyribonuclease I, Bovine Pancreas, Purified, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
Description
General Description
- Deoxyribonuclease I is an endonuclease that acts on phosphodiester bonds adjacent to pyrimidines to produce polynucleotides with terminal 5'-phosphates. In the presence of Mg2+, this compound cleaves each strand of DNA independently
Application
- In biomedical research, this compound is used to nick DNA as a first step to integrate labeled bases into DNA
- It can be used to eliminate genomic DNA from RNA solutions before RT-PCR
- It is also used during isolation and purification of plasma membrane vesicles from Neurospora crassa cell culture
- It can decrease viscosity in cell culture lysates, providing better yields in primary cell isolation
- It is also commonly used in combination with trypsin for the preparation of single cell suspension from rats
- Deoxyribonuclease I, bovine pancreas has been used in some studies in order to compare different methods for reducing RNase contamination in preparations of DNase
- This compound can be applied in studies to investigate the impact of the composition of sodium dodecyl sulfate (SDS) preparations on the renaturation of enzymes after polyacrylamide gel electrophoresis.

Specifications
Specifications
Quantity | 100 mg |
Form | Lyophilized |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction