Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEP-1/CD148 Antibody (261922), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB349460 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349460 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349460 Supplier Novus Biologicals Supplier No. FAB19341MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DEP-1/CD148 Monoclonal specifically detects DEP-1/CD148 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen DEP-1/CD148
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 261922
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of
Host Species Mouse
Immunogen E. coli-derived recombinant human DEP-1/CD148, Arg997-Ala1337, Accession # Q12913
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5795
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.