Learn More
Description
Specifications
Specifications
| Antigen | DEP-1/CD148 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM-CD148/05 |
| Conjugate | DyLight 405 |
| Dilution | Flow Cytometry |
| Gene Alias | CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of |
| Gene Symbols | PTPRJ |
| Host Species | Mouse |
| Immunogen | Human recombinant CD148 (amino acids 1-444). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
