Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEP-1/CD148 Antibody (MEM-CD148/05), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP221790G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221790G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221790G Supplier Novus Biologicals Supplier No. NBP221790G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DEP-1/CD148 Monoclonal specifically detects DEP-1/CD148 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen DEP-1/CD148
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-CD148/05
Conjugate DyLight 488
Dilution Flow Cytometry
Gene Alias CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of
Gene Symbols PTPRJ
Host Species Mouse
Immunogen Human recombinant CD148 (amino acids 1-444).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5795
Test Specificity The mouse monoclonal antibody MEM-CD148/05 recognizes CD148, a highly glycosylated up to 250 kDa receptor-like protein tyrosin phosphatase expressed mainly in lymphocytes, myeloid cells and epithelial cells.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.