Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEPDC7 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB289952 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289952 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289952 Supplier Novus Biologicals Supplier No. NBP232141PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEPDC7 Polyclonal antibody specifically detects DEPDC7 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen DEPDC7
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias DEP domain containing 7, DEP domain-containing protein 7, dJ85M6.4, dJ85M6.4 (novel 58.3 KDA protein), novel 58.3 KDA protein, Protein TR2/D15, TR2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 461 to 511 of human DEP Domain Containing 7 using the numbering given in entry NP_001070710.1 (GeneID 91614).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91614
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.