Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEPTOR/DEPDC6 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP149674A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP149674A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP149674A70 Supplier Novus Biologicals Supplier No. NBP149674AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEPTOR/DEPDC6 Polyclonal specifically detects DEPTOR/DEPDC6 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DEPTOR/DEPDC6
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DEP domain containing 6, DEP domain containing mTOR interacting protein, DEP domain containing MTOR-interacting protein, DEP domain-containing mTOR-interacting protein, DEP domain-containing protein 6, DEP.6, DEPDC6FLJ12341, DKFZp564B1778, FLJ12428, FLJ13854
Gene Symbols DEPTOR
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human DEPTOR protein (within residues 10-200). [Swiss-Prot: Q8TB45]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 64798
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.