Learn More
Description
Specifications
Specifications
| Antigen | DEPTOR/DEPDC6 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | DEP domain containing 6, DEP domain containing mTOR interacting protein, DEP domain containing MTOR-interacting protein, DEP domain-containing mTOR-interacting protein, DEP domain-containing protein 6, DEP.6, DEPDC6FLJ12341, DKFZp564B1778, FLJ12428, FLJ13854 |
| Gene Symbols | DEPTOR |
| Host Species | Rabbit |
| Immunogen | A genomic peptide made to an internal region of the human DEPTOR protein (within residues 10-200). [Swiss-Prot: Q8TB45] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
