Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEPTOR/DEPDC6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227083SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227083SS 0.025 mg
NBP227083 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227083SS Supplier Novus Biologicals Supplier No. NBP227083SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEPTOR/DEPDC6 Polyclonal specifically detects DEPTOR/DEPDC6 in Human, Rat, Equine, Primate samples. It is validated for Western Blot.

Specifications

Antigen DEPTOR/DEPDC6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3 - 5 ug/mL
Formulation PBS, 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q8TB45
Gene Alias DEP domain containing 6, DEP domain containing mTOR interacting protein, DEP domain containing MTOR-interacting protein, DEP domain-containing mTOR-interacting protein, DEP domain-containing protein 6, DEP.6, DEPDC6FLJ12341, DKFZp564B1778, FLJ12428, FLJ13854
Gene Symbols DEPTOR
Host Species Rabbit
Immunogen A portion of amino acids 230-300 of human DEPOR/DEPDC6 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64798
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee, horse and rat 93% homologous in mouse andcow, and 87% homologous in chicken.
Target Species Human, Rat, Equine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.