Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Derlin 1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB100448UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100448UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100448UV Supplier Novus Biologicals Supplier No. NB100448UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Derlin 1 Polyclonal specifically detects Derlin 1 in Canine, Chinese Hamster, Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Derlin 1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DER-1, DER1Degradation in endoplasmic reticulum protein 1, Der1-like domain family, member 1, Der1-like protein 1, derlin-1, DERtrin-1, FLJ13784, FLJ42092, MGC3067, PRO2577
Gene Symbols DERL1
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human Derlin 1 (between residues 200-251). [UniProtNo. Q9BUN8]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79139
Target Species Canine, Chinese Hamster, Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.