Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Derlin 1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB100448G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100448G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100448G Supplier Novus Biologicals Supplier No. NB100448G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Derlin 1 Polyclonal specifically detects Derlin 1 in Canine, Chinese Hamster, Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Derlin 1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9BUN8
Gene Alias DER-1, DER1Degradation in endoplasmic reticulum protein 1, Der1-like domain family, member 1, Der1-like protein 1, derlin-1, DERtrin-1, FLJ13784, FLJ42092, MGC3067, PRO2577
Gene Symbols DERL1
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human Derlin 1 (between residues 200-251). [UniProt# Q9BUN8]
Molecular Weight of Antigen 29 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ER Markers, Immunology, Protein Turnover, Signal Transduction, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 79139
Test Specificity Human, Canine and Chinese Hamster. Predicted to react with Xenopus, bovine, chicken, rat, zebrafish, and mouse based on 100% sequence homology. Mouse reactivity reported in scientific literature (PMID: 25377857).
Target Species Canine, Chinese Hamster, Canine, Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.