Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Derlin-2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB127247 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB127247 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB127247 Supplier Novus Biologicals Supplier No. NBP298598AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Derlin-2 Polyclonal specifically detects Derlin-2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Derlin-2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias carcinoma related, Degradation in endoplasmic reticulum protein 2, Der1-like domain family, member 2, Der1-like protein 2, DER2, derlin-2, F-LAN-1DERtrin-2, FLANa, F-LANaCGI-101
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Derlin-2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51009
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.