Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Derlin-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | Derlin-3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159466
![]() |
Novus Biologicals
NBP159466 |
100 μL |
Each for $480.74
|
|
|||||
NBP15946620
![]() |
Novus Biologicals
NBP15946620UL |
20 μL | N/A | N/A | N/A | ||||
Description
Derlin-3 Polyclonal specifically detects Derlin-3 in Human samples. It is validated for Western Blot.Specifications
| Derlin-3 | |
| Polyclonal | |
| Rabbit | |
| Q96Q80 | |
| 91319 | |
| Synthetic peptides corresponding to Derlin 3(Der1-like domain family, member 3) The peptide sequence was selected from the middle region of Derlin 3. Peptide sequence FFFNMLFVFRYCRMLEEGSFRGRTADFVFMFLFGGVLMTLLGLLGSLFFL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| C22orf14, chromosome 22 open reading frame 14, Degradation in endoplasmic reticulum protein 3, Der1-like domain family, member 3, Der1-like protein 3, DER3, derlin 3, derlin-3, DERtrin 3, DERtrin-3, FLJ43842, IZP6, LLN2, MGC71803 | |
| DERL3 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title