Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Derlin-3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15946620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15946620 20 μL
NBP159466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15946620 Supplier Novus Biologicals Supplier No. NBP15946620UL

Rabbit Polyclonal Antibody

Derlin-3 Polyclonal specifically detects Derlin-3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Derlin-3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96Q80
Gene Alias C22orf14, chromosome 22 open reading frame 14, Degradation in endoplasmic reticulum protein 3, Der1-like domain family, member 3, Der1-like protein 3, DER3, derlin 3, derlin-3, DERtrin 3, DERtrin-3, FLJ43842, IZP6, LLN2, MGC71803
Gene Symbols DERL3
Host Species Rabbit
Immunogen Synthetic peptides corresponding to Derlin 3(Der1-like domain family, member 3) The peptide sequence was selected from the middle region of Derlin 3. Peptide sequence FFFNMLFVFRYCRMLEEGSFRGRTADFVFMFLFGGVLMTLLGLLGSLFFL.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91319
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.