Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dermcidin Mouse anti-Human, Clone: G-81, Novus Biologicals™
SDP

Catalog No. NB018651 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018651 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB018651 Supplier Novus Biologicals Supplier No. NBP1496200.25ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Dermcidin Monoclonal specifically detects Dermcidin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoassay.

Specifications

Antigen Dermcidin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone G-81
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay
Formulation No buffer with 0.02% Sodium Azide
Gene Accession No. P81605
Gene Alias AIDDdiffusible survival/evasion peptide, DCD-1, dermcidin, DSEPpreproteolysin, HCAP, MGC71930, PIF, Preproteolysin, proteolysis inducing factor, survival promoting peptide
Gene Symbols DCD
Host Species Mouse
Immunogen Fractioned sweat peptides in complete Freunds adjuvant
Purification Method Tissue culture supernatant
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117159
Test Specificity The monoclonal antibody G-81 was produced against a sweat-specific antigen. It reacts to sweat but not to other body fluids (e.g. serum, saliva, semen, milk, urine and tears) in ELISA. The N-terminal amino acid sequence (18 amino acids residues) is identical to a part of human cachexia-associated protein and of a survival-promoting peptide for neuronal cells. Besides this, the sequence is identical to the C-terminal peptide (amino acids 62-79) of DCD. The monoclonal antibody G81 can be used as a specific marker for eccrine glands in the human skin.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.