Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dermcidin Mouse anti-Human, Clone: G-81, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$243.50
Specifications
Antigen | Dermcidin |
---|---|
Clone | G-81 |
Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
Dermcidin Monoclonal specifically detects Dermcidin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoassay.Specifications
Dermcidin | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay | |
Monoclonal | |
Supernatant | |
RUO | |
P81605 | |
117159 | |
Fractioned sweat peptides in complete Freunds adjuvant | |
Primary | |
Store at 4C. Do not freeze. |
G-81 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
AIDDdiffusible survival/evasion peptide, DCD-1, dermcidin, DSEPpreproteolysin, HCAP, MGC71930, PIF, Preproteolysin, proteolysis inducing factor, survival promoting peptide | |
DCD | |
IgM | |
Tissue culture supernatant | |
The monoclonal antibody G-81 was produced against a sweat-specific antigen. It reacts to sweat but not to other body fluids (e.g. serum, saliva, semen, milk, urine and tears) in ELISA. The N-terminal amino acid sequence (18 amino acids residues) is identical to a part of human cachexia-associated protein and of a survival-promoting peptide for neuronal cells. Besides this, the sequence is identical to the C-terminal peptide (amino acids 62-79) of DCD. The monoclonal antibody G81 can be used as a specific marker for eccrine glands in the human skin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title