Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DERP1 Antibody (H.AA.IV7A3), Novus Biologicals™
SDP

Catalog No. DDX0211P100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0211P100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0211P100 Supplier Novus Biologicals Supplier No. DDX0211P100
Add to Cart
Add to Cart

Human Monoclonal Antibody has been used in 2 publications

DERP1 Monoclonal specifically detects DERP1 in Insect samples. It is validated for ELISA, ELISA Capture (Matched Antibody Pair), Western Blot.

Specifications

Antigen DERP1
Applications ELISA, ELISA Capture (Matched Antibody Pair), Western Blot
Classification Monoclonal
Clone H.AA.IV7A3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, ELISA Capture (Matched Antibody Pair) 1:100-1:2000
Gene Alias Der p 1 allergen, DERP-1
Host Species Human
Immunogen Derp-1 was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Insect
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.