Learn More
Description
Specifically detects Desert Hedgehog/Dhh in Human samples, and it is validated for Western Blotting
Specifications
Specifications
| Antigen | Desert Hedgehog/Dhh |
| Applications | Western Blot, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000-1:20000, Immunohistochemistry, Immunohistochemistry-Frozen |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal |
| Gene Alias | desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, HHG-3MGC35145 |
| Gene Symbols | DHH |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of human Dhh. The exact sequence is proprietary. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
