Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Desformylflustrabromine hydrochloride
GREENER_CHOICESDP

Catalog No. 332810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-281-0 10 mg
33-285-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-281-0 Supplier Tocris Bioscience™ Supplier No. 3328/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Positive allosteric modulator of α4β2; also muscle-type nAChR inhibitor

Positive allosteric modulator of nicotinic α4β2 receptors; selectively increases the ionic current through α4β2 in the presence of ACh. Displays 14.7-fold selectivity for α4β2 over homomeric (α7) receptors. Moderately cytotoxic in HCT-116 cells. Also inhibits human muscle (αβεδ) and Torpedo (αβγδ) nAChRs (IC50 values are 1.0 and 0.1 μM, respectively) by binding in the ion channel.

Chemical Identifiers

CAS 951322-11-5
Molecular Formula C16H22BrClN2

Specifications

Quantity 10 mg
Formula Weight 357.72
Purity >98%
Molecular Weight (g/mol) 357.72
Chemical Name or Material Desformylflustrabromine hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.