Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DESI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562786 Supplier Invitrogen™ Supplier No. PA562786
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IMLGKQLEGI WHTSIVVHKD EFFFGSGGIS SCPPGGTLLG PPDSVVDVGS TEVTEE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Protease which deconjugates SUMO1, SUMO2 and SUMO3 from some substrate proteins. Has isopeptidase but not SUMO-processing activity. Desumoylates ZBTB46. Collaborates with UBQLN4 in the export of ubiquitinated proteins from the nucleus to the cytoplasm (PubMed:29666234). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DESI1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DESI1
Gene Accession No. Q6ICB0
Gene Alias D15Wsu75e; DESI1; DeSI-1; DESI2; desumoylating isopeptidase 1; desumoylating isopeptidase 2; DJ347H13.4; FAM152B; family with sequence similarity 152, member B; Polyubiquitinated substrate transporter; POST; PPPDE peptidase domain containing 2; PPPDE peptidase domain-containing protein 2; PPPDE2
Gene Symbols DESI1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DESI1. Recombinant protein control fragment (Product #RP-100920).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27351
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.