Learn More
Kemtec™ Designing Investigation in Blood Spatter Kit
SCI_ED
Catalog No. S27540 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
9 to 14
Catalog No. Grade
S27540 9 to 14
1 options
Catalog No. S27540 Supplier Kemtec™ Supplier No. 10552
Kemtec™ Designing Investigation in Blood Spatter Kit guides students through the investigative process of a blood spatter detective with an inquiry based activity.
  • Students design and conduct a simple experiment to model how blood reacts and how blood spatter patterns change in response to variables.
  • Student may choose to investigate the effects of angle, force, distance, surface texture, rate of movement, velocity at impact, number of blows, or location of the victim to test a theory.
  • Students may choose to investigate the difference in spatter due to cast-off, blunt force trauma, or projectile trauma or to compare patterns due to impact spatter, passive spatter, transfer spatter, or projected spatter.
  • Just like the detective analyzes and presents their results as evidence to support a case, the student presents their hypothesis, experimental design, variables, results and conclusion in a Power Point Presentation.
  • Includes an instructor's manual with reproducible student pages.
  • May be used independently or as an introduction or companion to the analysis activities provided in Kemtec's Blood Spatter Kit.
  • Meets National Standards for Grade Levels 9 to 14.
Specifications
Grade 9 to 14
Age 14 to 20
Includes Instructor's Manual, , Simulated Blood, Hammer, Roll of Butcher Paper, Scissors, Plastic knives, Screw Drivers, Protractors, Retractable Tape measures, Plastic Pipets
Weight (English) 7 lb.
Dimensions (L x W x H) 20 x 12 x 5 in.