Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Antibody (D33), Novus Biologicals™
SDP

Catalog No. NB12017156 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12017156 1 mL
NB018085 0.1 mL
NB120171565 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB12017156 Supplier Novus Biologicals Supplier No. NB12017156
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen Desmin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone D33
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Gene Alias CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
Gene Symbols DES
Host Species Mouse
Immunogen BALB/C mice were immunized with purified desmin from human muscle.
Purification Method Tissue culture supernatant
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 1674
Test Specificity This antibody reacts with desmin protein of 53 kDa. Both striated as well as smooth muscle cells stain well with this antibody. The antibody shows a broad interspecific cross-reactivity, reacting with human.
Target Species Human, Rat, Chicken
Content And Storage Store at 4°C.
Form Supernatant
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.