Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Antibody (DES/2960R), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB385226 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385226 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385226 Supplier Novus Biologicals Supplier No. NBP308664FR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Desmin Monoclonal antibody specifically detects Desmin in Human, Rat, Chicken, Hamster samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Desmin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DES/2960R
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
Host Species Rabbit
Immunogen Recombinant human full-length Desmin protein (Uniprot: P17661)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 1674
Target Species Human, Rat, Chicken, Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.