Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Antibody (OTI4G1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP270565FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270565FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270565FR Supplier Novus Biologicals Supplier No. NBP270565FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Desmin Monoclonal specifically detects Desmin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Desmin
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI4G1
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
Gene Symbols DES
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 104-391 of human Desmin (NP_001918) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cytoskeleton Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1674.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.