Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Desmin (Muscle Cell Marker) Ab-1, Mouse Monoclonal Antibody

Specifically detect desmin in human, baboon, monkey, cow, cat, dog, hamster, rat and chicken samples.

$264.00 - $909.00

Specifications

Antigen Desmin (Muscle Cell Marker) Ab-1
Clone D33
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS376S0
View Documents
Epredia™
MS376S0
Concentrated 100 μL
Each for $264.00
Only null left
Add to Cart
 
MS376S1
View Documents
Epredia™
MS376S1
Concentrated 500 μL
Each for $671.00
Only null left
Add to Cart
 
MS376S
View Documents
Epredia™
MS376S
Concentrated 1 mL
Each for $909.00
Only null left
Add to Cart
 
MS376R7
View Documents
Epredia™
MS376R7
Prediluted 7 mL
Each for $380.00
Only null left
Add to Cart
 
MS376RQ
View Documents
Epredia™
MS376RQ
Ready-to-Use 12 mL
Each for $547.50
Only null left
Add to Cart
 
MS376R1
View Documents
Epredia™
MS376R1
Prediluted 1 mL N/A N/A N/A
Description

Description

Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin. It reacts with leiomyosarcomas (smooth muscle) as well as rhabdomyosarcomas (striated muscle).

Host Species: Mouse

Clone: D33

Isotype: IgG1/κ

Species Reactivity: Human, Baboon, Monkey, Cow, Cat, Dog, Hamster, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Desmin from human muscle

Molecular Weight: 53 kDa

Positive Control: Muscle or Sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Specifications

Desmin (Muscle Cell Marker) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Desmin from human muscle
D33
Monoclonal
Mouse
Muscle Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.