Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmocollin-1 Antibody (772906) [DyLight 488], Novus Biologicals™
SDP

Catalog No. NB374754 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374754 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374754 Supplier Novus Biologicals Supplier No. FAB7367K
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Desmocollin-1 Monoclonal antibody specifically detects Desmocollin-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF

Specifications

Antigen Desmocollin-1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 772906
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, desmosomal glycoprotein 2/3, DG2/DG3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Desmocollin-1, Arg135-Lys691, Accession # P55849
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1823
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.