Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmocollin-2 Antibody (003), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB041304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB041304 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB041304 Supplier Novus Biologicals Supplier No. NBP290904AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Desmocollin-2 Monoclonal specifically detects Desmocollin-2 in Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Desmocollin-2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ARVD11, Cadherin family member 2, CDHF2DGII/III, desmocollin 2, desmocollin-2, Desmocollin-3, Desmosomal glycoprotein II, desmosomal glycoprotein II/III, Desmosomal glycoprotein III, DG2, DSC3DKFZp686I11137
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat Desmocollin-2 (Q3T1K6; Met1-Pro694).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1824
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.