Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmocollin-2 Antibody (7G6), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N254519APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N254519APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N254519APC7 Supplier Novus Biologicals Supplier No. NBP254519APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Desmocollin-2 Monoclonal specifically detects Desmocollin-2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Desmocollin-2
Applications Flow Cytometry
Classification Monoclonal
Clone 7G6
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ARVD11, Cadherin family member 2, CDHF2DGII/III, desmocollin 2, desmocollin-2, Desmocollin-3, Desmosomal glycoprotein II, desmosomal glycoprotein II/III, Desmosomal glycoprotein III, DG2, DSC3DKFZp686I11137
Gene Symbols DSC2
Host Species Mouse
Immunogen Human desmocollin-2 extracellular domain (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1824.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.