Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

desmosome/hemidesmosome Human anti-Human, Clone: F12, Novus Biologicals™
SDP

Catalog No. NB141995 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB141995 0.05 mg
NB413780 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB141995 Supplier Novus Biologicals Supplier No. NBP2812790.05MG
Add to Cart
Add to Cart

Human Monoclonal Antibody

desmosome/hemidesmosome Monoclonal specifically detects desmosome/hemidesmosome in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Electron Microscopy.

Specifications

Antigen desmosome/hemidesmosome
Applications Flow Cytometry, Immunofluorescence, Immunomicroscopy
Classification Monoclonal
Clone F12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence, Electron Microscopy
Formulation PBS
Host Species Human
Immunogen F12 was established by fusing peripheral blood lymphocytes derived from a human vulgaris patient with a hetermyeloma cell line.
Purification Method Affinity purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.